• USA Home
  • D6531 - Dimethyl-DL-2,3-distearoyloxypropyl(2-hydroxyethyl)ammonium acetate

EMAIL THIS PAGE TO A FRIEND
D6531 Sigma

Dimethyl-DL-2,3-distearoyloxypropyl(2-hydroxyethyl)ammonium acetate

  • CAS Number 13106-14-4

  • Empirical Formula (Hill Notation) C43H86NO5

  • Molecular Weight 697.15

  •  MDL number MFCD00036853

Purchase

Properties

Related Categories Alcohol Acetates, Biochemicals and Reagents, Branched fatty acids, Fatty Acyls, Fatty Esters,
InChI Key   DRUSFJBLMPJNIZ-UHFFFAOYSA-M
storage temp.   −20°C

Description

Biochem/physiol Actions

Stimulator of HeLa cell alkaline phosphatase.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?