• USA Home
  • D6562 - Anti-DP-1 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
D6562 Sigma

Anti-DP-1 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Gene Regulation, Antibodies to Common Transcription Factors, Antibodies to Transcription Factors,
species reactivity   human
application(s)   indirect immunofluorescence: 1:50 using DP-1 transfected COS7 cells
  western blot: 1:3,000 using human recombinant DP-1 in vitro translated or expressed in COS7 cells
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt 50 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... TFDP1(7027)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the N-terminal of human DP-1, amino acids 9-27 with C-terminally added cysteine, conjugated to KLH.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Specificity

The epitope recognized by the antibody is conserved in mouse.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?