• USA Home
  • D6629 - 2,4-Dinitro-5-fluoroacetanilide

EMAIL THIS PAGE TO A FRIEND
D6629 Sigma

2,4-Dinitro-5-fluoroacetanilide

  • CAS Number 1548-18-1

  • Empirical Formula (Hill Notation) C8H6FN3O5

  • Molecular Weight 243.15

  •  MDL number MFCD00047669

  •  PubChem Substance ID 24894039

Purchase

Properties

Related Categories Biochemicals and Reagents, Luminescent Compounds/Detection, Other luminescent compounds More...
InChI Key   LZBXUWOTXPNTNQ-UHFFFAOYSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?