• USA Home
  • D6678 - 3,5-Dinitrobenzyl-N-(propyl)amine hydrochloride

EMAIL THIS PAGE TO A FRIEND
D6678 Sigma

3,5-Dinitrobenzyl-N-(propyl)amine hydrochloride

≥98%

Synonym: DNBPA

  • CAS Number 127312-05-4

  • Empirical Formula (Hill Notation) C10H13N3O4 · HCl

  • Molecular Weight 275.69

  •  MDL number MFCD00274419

Purchase

Properties

Related Categories Amine Salts, Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   HVOKZJCNBLTMRL-UHFFFAOYSA-N
assay   ≥98%
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?