• USA Home
  • D6782 - DMT-Thymidine H-phosphonate triethylammonium salt

EMAIL THIS PAGE TO A FRIEND
D6782 Sigma

DMT-Thymidine H-phosphonate triethylammonium salt

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Nucleotide Analogs More...
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?