• USA Home
  • D7013 - 1,2-Dipalmitoyl-sn-glycerol 3-phosphate diphenyl ester

EMAIL THIS PAGE TO A FRIEND
D7013 Sigma

1,2-Dipalmitoyl-sn-glycerol 3-phosphate diphenyl ester

~99%

Synonym: 1,2-Dihexadecanoyl-sn-glycero-3-(bis[phenyl] phosphate)

Purchase

Properties

Related Categories Biochemicals and Reagents, Glycerophosphates, Glycerophospholipids, Lipids
InChI Key   XDTOUWPJZGEUQR-GWHBCOKCSA-N
assay   ~99%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?