• USA Home
  • D7125 - Deoxycorticosterone 21-glucoside

EMAIL THIS PAGE TO A FRIEND
D7125 Sigma

Deoxycorticosterone 21-glucoside

Synonym: 21-Hydroxy-4-pregnene-3,20-dione 21-glucoside, 21-Hydroxyprogesterone 21-glucoside, 4-Pregnen-21-ol-3,20-dione 21-glucoside, Cortexone 21-glucoside

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Corticosteroid, Cytokines, Growth Factors and Hormones, Hormones,
InChI Key   SMSMUZPFFJLROV-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?