EMAIL THIS PAGE TO A FRIEND
D7275 Sigma

Disopyramide

Synonym: α-Diisopropylaminoethyl-α-phenylpyridine-2-acetamide

  • CAS Number 3737-09-5

  • Empirical Formula (Hill Notation) C21H29N3O

  • Molecular Weight 339.47

  •  MDL number MFCD00057366

Purchase

Properties

InChI Key   UVTNFZQICZKOEM-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Biochem/physiol Actions

Class IA antiarrhythmic; sodium channel blocker

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?