EMAIL THIS PAGE TO A FRIEND
D7285 Sigma

3′-Deoxyguanosine

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   OROIAVZITJBGSM-UHFFFAOYSA-N

Description

Application

3′-Deoxyguanosine (3dG) is used as a complexing ligand for enzymes, such as purine nucleoside phosphorylase (PNP), and receptors, such as G-coupled receptors, to enable structure analysis. 3′-Deoxyguanosine may be used to selectively impair transcription but not polyadenylation.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?