EMAIL THIS PAGE TO A FRIEND
D7389 Sigma

Tiron

Synonym: 4,5-Dihydroxy-1,3-benzenedisulfonic acid disodium salt, Pyrocatechol-3,5-disulfonic acid disodium salt

  • CAS Number 149-45-1

  • Empirical Formula (Hill Notation) C6H4Na2O8S2

  • Molecular Weight 314.20

  •  Beilstein Registry Number 4629740

  •  EC Number 205-741-5

  •  MDL number MFCD00007473

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

InChI Key   ISWQCIVKKSOKNN-UHFFFAOYSA-L

Description

Application

Cell permeable superoxide scavenger.

Other Notes

This product has been replaced by 172553-ALDRICH | 4,5-Dihydroxy-1,3-benzenedisulfonic acid disodium salt monohydrate, | (OH)2C6H2(SO3Na)2

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

172553 4,5-Dihydroxy-1,3-benzenedisulfonic acid disodium salt monohydrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?