• USA Home
  • D7416 - Duolite® weakly acidic cation exchanger Cross-linked polyacrylic acid

EMAIL THIS PAGE TO A FRIEND
D7416 Sigma

Duolite® weakly acidic cation exchanger Cross-linked polyacrylic acid

16-50 mesh (wet)

Purchase

Properties

contains   ~50% moisture content
particle size   16-50 mesh (wet)

Description

Other Notes

C-433, Gel form
Ionic form: Hydrogen
Maximum operating temperature: 120 °C
Effective pH range: 5-14
Capacity: 2 meq/ml; 5.5 meq/g

Legal Information

Duolite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

546976 Dowex® MAC-3 hydrogen form, hydrogen form

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?