• USA Home
  • D7544 - [Glu52]-Diphtheria toxin from Corynebacterium diphtheriae

EMAIL THIS PAGE TO A FRIEND
D7544 Sigma

[Glu52]-Diphtheria toxin from Corynebacterium diphtheriae

lyophilized powder

Synonym: CRM 197

Purchase

Properties

form   lyophilized powder
storage temp.   2-8°C

Description

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing sodium phosphate and lactose

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

D2189 [Glu52]-Diphtheria toxin from Corynebacterium diphtheriae, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?