• USA Home
  • D7632 - 2′,3′-Di-O-benzoyluridine

EMAIL THIS PAGE TO A FRIEND
D7632 Sigma

2′,3′-Di-O-benzoyluridine

≥99% (TLC)

Synonym: Uridine 2′,3′-dibenzoate

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   FZFFWKZFQMIXKC-FESZSOIMSA-N
assay   ≥99% (TLC)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?