• USA Home
  • D7754 - DNP-ε-amino-n-caproic acid

EMAIL THIS PAGE TO A FRIEND
D7754 Sigma

DNP-ε-amino-n-caproic acid

Synonym: N-(2,4-Dinitrophenyl)-ε-amino-n-caproic acid

Purchase

Properties

Related Categories Amino Acid Analysis, Amino Acid Analysis Reagents, Biochemicals and Reagents, Peptide Analysis and Characterization, Peptides More...
InChI Key   ZYUWUKIAUDIXCQ-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?