• USA Home
  • D8300 - N,N-Dimethylformamide

EMAIL THIS PAGE TO A FRIEND
D8300 Sigma

N,N-Dimethylformamide

suitable for neutral marker for measuring electroosmotic flow (EOF), ~99%

Synonym: DMF, NSC 5356

Purchase

Properties

Related Categories Dimethylformamide (DMF), Solvent by Type, Solvents More...
vapor density   2.5 (vs air)
vapor pressure   2.7 mmHg ( 20 °C)
InChI Key   ZMXDDKWLCZADIW-UHFFFAOYSA-N
assay   ~99%
form   liquid
autoignition temp.   833 °F
expl. lim.   15.2 %
refractive index   n20/D 1.430(lit.)
bp   153 °C(lit.)
mp   −61 °C(lit.)
density   0.944 g/mL(lit.)
suitability   suitable for neutral marker for measuring electroosmotic flow (EOF)

Description

Application

Solvent for many hydrophobic organic compounds.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2265 3 / PGIII
WGK Germany 
2
RTECS 
LQ2100000
Flash Point(F) 
136.4 °F
Flash Point(C) 
58 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?