• USA Home
  • D8554 - Nunclon® cell culture dishes

EMAIL THIS PAGE TO A FRIEND
D8554 Sigma

Nunclon® cell culture dishes

diam. × H 150 mm × 20 mm

Synonym: cell culture dish, cell culture plate, cell culture plates, petri dishes

Purchase

Properties

Related Categories Cell Culture Supplies, Culture Dishes, Labware, Manufacturer Browser, Microbiology Equipment,
material   polystyrene dishes with lids
  round
description   round, vented
sterility   sterile; γ-irradiated
packaging   case of 80
mfr. no.   Nunc
diam. × H   150 mm × 20 mm
surface area   145 cm^2
matrix active group   Nunclon surface treatment

Description

General description

• Large range available, treated for cell culture but with many other uses
• Optically clear and very flat for use in microscope
• Non-toxic
• Certified surface treatment for optimal cell attachment and growth

Legal Information

Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?