D8562 Sigma
Dispo-Biodialyzer
MWCO 0.1 kDa
DOWNLOAD MSDS (PDF)
Properties
pore size   0.1 kDa MWCO
Description
General description
A unique disposable unit for small sample dialysis, 5-100 μL. Dialysis is rapid with the ultra thin membrane of regenerated cellulose. After dialysis, sample can be recovered, almost 100% by centrifugation in a microcentrifuge tube. Recommended for salt removal, buffer exchange, protein binding assays and as a equilibrium dialyzer.
Price and Availability
Customers Also Viewed
Biodialyser system
single sided, 50 μL
Cellulose acetate membrane
dialyser capacity 10-200 μL, MWCO 500 Da, diam. 11 mm
Cellulose acetate membrane
dialyser capacity 500-1,500 μL, MWCO 500 Da, diam. 16 mm
Avg. flat width 25 mm (1.0 in.)
avg. flat width 10 mm (0.4 in.)
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?