• USA Home
  • D8666 - DNA Spooling Educational Kit

EMAIL THIS PAGE TO A FRIEND
D8666 Sigma

DNA Spooling Educational Kit

Purchase

Properties

usage   1 kit sufficient for 25 students
storage temp.   −20°C

Description

Application

Kit provides "hands on" demonstration of DNA spooling (precipitation).

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?