• USA Home
  • D8775 - 4-Dimethylaminoazobenzene 4′-isothiocyanate

EMAIL THIS PAGE TO A FRIEND
D8775 Sigma

4-Dimethylaminoazobenzene 4′-isothiocyanate

Synonym: DABITC

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?