EMAIL THIS PAGE TO A FRIEND
D8937 Sigma
Dispo-Biodialyzer
MWCO 5 kDa
Purchase
Properties
Related Categories Biodialyser, Dialysis, Labware More...
pore size   5 kDa MWCO
Description
General description
A unique disposable unit for small sample dialysis, 5-100 μL. Dialysis is rapid with the ultra thin membrane of regenerated cellulose. After dialysis, sample can be recovered, almost 100% by centrifugation in a microcentrifuge tube. Recommended for salt removal, buffer exchange, protein binding assays and as a equilibrium dialyzer.
The Micro DispoDIALYZER includes a membrane, foam float, cap and two collection tubes (1.5 mL).
Price and Availability
Centricon Filter Units
Mitex PTFE Membrane Filters
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?