• USA Home
  • D9039 - Anti-Dideoxyinosine antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
D9039 Sigma

Anti-Dideoxyinosine antibody produced in rabbit

whole antiserum, lyophilized powder

Synonym: Anti-ddI

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, DE-DL, Primary Antibodies
biological source   rabbit
antibody form   whole antiserum
clone   polyclonal
form   lyophilized powder
usage   sufficient for ≥100 test
application(s)   radioimmunoassay: suitable using 3H-ddI
conjugate   unconjugated
storage temp.   2-8°C

Description

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Dideoxyinosine (2′,3′-dideoxyinosine, ddI, didanosine) is an inosine analog that has been shown to be effective in inhibiting retroviral activity.

Immunogen

ddI-BSA

Legal Information

U.S. Patent No. 5,221,611

Specificity

Anti-ddI has low cross reactivity with inosine, dideoxyadenosine, adenosine, hypoxanthine, and uric acid.

Physical form

Lyophilized from 1 mL dialyzed antiserum and prediluted in 0.01 M phosphate buffer containing 1% lactose.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?