• USA Home
  • D9387 - Didansyl-1,3-diaminopropane

EMAIL THIS PAGE TO A FRIEND
D9387 Sigma

Didansyl-1,3-diaminopropane

powder

  • CAS Number 64144-62-3

  • Empirical Formula (Hill Notation) C27H32N4O4S2

  • Molecular Weight 540.70

  •  MDL number MFCD00056583

  •  PubChem Substance ID 24894289

Purchase

Properties

Related Categories A - H, Amino Acids, Amino acids and other organic acid conjugates, Biochemicals and Reagents, Fluorescent Conjugates,
InChI Key   GVQXAPGVWXVSSZ-UHFFFAOYSA-N
form   powder
color   yellow to green
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?