• USA Home
  • D9545 - (5S,6S)-Dihydroxy-7-(mesitylenesulfonyloxy)heptanoic acid ethyl ester

EMAIL THIS PAGE TO A FRIEND
D9545 Sigma

(5S,6S)-Dihydroxy-7-(mesitylenesulfonyloxy)heptanoic acid ethyl ester

≥98% (HPLC)

Purchase

Properties

assay   ≥98% (HPLC)

Description

Application

Intermediate in the synthesis of leukotrienes

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?