• USA Home
  • D9751 - 2,4-Diamino-6-hydroxypyrimidine

EMAIL THIS PAGE TO A FRIEND
D9751 Sigma

2,4-Diamino-6-hydroxypyrimidine

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D19206 2,4-Diamino-6-hydroxypyrimidine, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?