• USA Home
  • DNP25 - L-Amino acids, dinitrophenyl

EMAIL THIS PAGE TO A FRIEND
DNP25 Sigma

L-Amino acids, dinitrophenyl

Synonym: (DNP)

Purchase

Properties

storage temp.   −20°C

Description

Packaging

25 2,4-Dinitrophenyl L-Amino Acids

Price and Availability

Standard component

Description

Product #

Add to Cart

N,S-diDNP-L-Cysteine 25 mg SDS    
N,N-di-DNP-L-Cystine 25 mg SDS    
N,N′-di-DNP-L-Histidine 25 mg SDS D9629
N,N′-di-DNP-L-Lysine 25 mg SDS    
N,N′-di-DNP-L-Ornithine 25 mg SDS    
N-DNP-L-Alanine 25 mg SDS    
N-DNP-L-Arginine 25 mg SDS D8129
N-DNP-L-Asparagine 25 mg SDS    
N-DNP-L-Aspartic Acid 25 mg SDS    
N-DNP-L-Glutamic Acid, Cyclohexylamine Salt 25 mg SDS D9254
N-DNP-Glycine 25 mg SDS D9504
N-DNP-L-Isoleucine 25 mg SDS    
N-DNP-L-Leucine 25 mg SDS    
Nε-DNP-L-Lysine Hydrochloride 25 mg SDS D0380
N-DNP-DL-Methionine 25 mg SDS    
N-DNP-DL-Norleucine 25 mg SDS    
N-DNP-DL-Norvaline 25 mg SDS D1005
Nδ-DNP-L-ORNITHINE Hydrochloride 25 mg SDS D1130
N-DNP-L-Phenylalanine 25 mg SDS    
N-DNP-L-Proline 25 mg SDS    
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?