• USA Home
  • E005 - Benzodiazepine Receptor

EMAIL THIS PAGE TO A FRIEND
E005 Sigma

Benzodiazepine Receptor

lyophilized powder

Purchase

Properties

form   lyophilized powder
usage   sufficient for 100 tests
storage temp.   2-8°C

Description

Reconstitution

400 mg receptor preparation (the contents of one vial) is suspended in 40 mL sodium phosphate buffer on the day of the assay. The suspension is stable for one day at RT and for more than 8 months at -80 °C. It can be thawed and refrozen several times during this period.

Application

The preparation contains a receptor binding protein in a stable, lyophilized form convenient to use for a radioreceptor assay (RRA) for benzodiazepines. The method is based on the competition for specific receptors between biologically active benzodiazepine in biological fluids and 3H-flunitrazepam. The specificity of the receptor preparation is similar to that of fresh membrane-bound rat brain benzodiazepine receptors. Lorazepam is used as a standard unless a specific benzodiazepne is preferred.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?