• USA Home
  • E0125 - Escherichia coli acetone powders from Escherichia coli ATCC 11246

EMAIL THIS PAGE TO A FRIEND
E0125 Sigma

Escherichia coli acetone powders from Escherichia coli ATCC 11246

Purchase

Properties

Related Categories Acetone Powders, Biochemicals and Reagents, Proteins and Derivatives More...
storage temp.   −20°C

Description

Application

Source of L-glutamic decarboxylase and L-asparaginase

Biochem/physiol Actions

Aprotinin is a competitive serine protease inhibitor that inhibits trypsin, chymotrypsin, kallikrein and plasmin. It is often used to suppress proteolysis during isolation of proteins from cells and tissues. Aprotinin forms a stable 1:1 complex with the protease and blocks its active site. Binding is reversible with most aprotinin-protease complexes dissociating at pH >10 or <3. Effective concentration equimolar with protease.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?