• USA Home
  • E0403 - Ephrin-A4/Fc Chimera human

EMAIL THIS PAGE TO A FRIEND
E0403 Sigma

Ephrin-A4/Fc Chimera human

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym: EFL-4, LERK-4

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cytokines and Growth Factors, Cytokines, Growth Factors and Hormones, Ephrin,
recombinant   expressed in NSO cells
assay   >90% (SDS-PAGE)
form   lyophilized powder
mol wt   monomer calculated mol wt 43.7 kDa
  mol wt ~50 kDa by SDS-PAGE (reducing)
impurities   endotoxin, tested
storage temp.   −20°C
Gene Information   human ... EFNA4(1945)

Description

Analysis Note

The biological activity is measured by its ability to bind immobilized recombinant mouse EphA7/Fc in ELISA.

Biochem/physiol Actions

Member of the ephrin ligand family that binds Eph receptor tyrosine kinases (EphA2, EphA3, EphA4, EphA5, EphA6, EphA7, and EphB1). Involved in pattern formation and morphogenesis.

Other Notes

Extracellular domain of human ephrin-A4 (amino acids 1-171) fused to the C-terminal 6X histidine-tagged Fc region of human IgG1.

Physical form

Lyophilized from a 0.2 μm filtered solution in 20 mM Tris and 0.2 M sodium chloride, pH 8.5

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?