• USA Home
  • E0510 - Eosin 5-isothiocyanate–dextran 70S

EMAIL THIS PAGE TO A FRIEND
E0510 Sigma

Eosin 5-isothiocyanate–dextran 70S

Synonym: EITC–Dextran

Purchase

Properties

mol wt   mol wt ~70,000
extent of labeling   ~0.002 mol EITC per mol glucose
storage temp.   2-8°C

Description

Application

Dextran labeled with eosin 5-isothiocyanate for use in perfusion studies in animals.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

R9379 Rhodamine B isothiocyanate–Dextran, average mol wt ~70,000

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?