EMAIL THIS PAGE TO A FRIEND
E0627 Sigma

d-Equilenin

Synonym: 1,3,5(10),6,8-Estrapentaen-3-ol-17-one, 3-Hydroxy-1,3,5(10),6,8-estrapentaen-17-one

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
KG6125000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?