• USA Home
  • E0762 - N-Acetyl-β-Endorphin Fragment 1-27 Human

EMAIL THIS PAGE TO A FRIEND
E0762 Sigma

N-Acetyl-β-Endorphin Fragment 1-27 Human

≥97% (HPLC)

  • CAS Number 76622-86-1

  • Empirical Formula (Hill Notation) C141H219N33O41S

  • Molecular Weight 3064.51

  •  MDL number MFCD00076391

  •  PubChem Substance ID 329798989

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Opioid Peptides, Other Opioid Peptides, Peptides and Proteins,
InChI Key   KBTYRLSAENUDCP-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Ac-Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-Leu-Phe-Lys-Asn-Ala-Ile-Ile-lys-Asn-Ala-Tyr

Other Notes

Predominant form of β-endorphin in the intermediate lobe of the pituitary under normal conditions

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?