• USA Home
  • E0782 - Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in rat

EMAIL THIS PAGE TO A FRIEND
E0782 Sigma

Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in rat

clone 176436, purified immunoglobulin, lyophilized powder

Synonym: Anti-Egfr, Anti-Erbb, Anti-Errp, Anti-Wa5

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Cytokines and Growth Factors, Antibodies to Epidermal Growth Factors (EGF) and NRG Family,
species reactivity   mouse
application(s)   flow cytometry: 100 μg/mL
  western blot: 1-2 μg/mL
clone   176436, monoclonal
antibody form   purified immunoglobulin
form   lyophilized powder
isotype   IgG1
storage temp.   −20°C
Gene Information   mouse ... Egfr(13649)
biological source   rat
conjugate   unconjugated

Description

Immunogen

purified, NSO cell-derived recombinant mouse epidermal growth factor receptor extracellular domain.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, type N95 (US), type P1 (EN143) respirator filter
WGK Germany  1
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

E2156 Monoclonal Anti-EGF Receptor antibody produced in mouse, clone 225, purified from hybridoma cell culture
HPA001200 Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
E1282 Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
E0532 Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 102618, purified immunoglobulin, lyophilized powder
E2760 Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 29.1, ascites fluid
E3641 Epidermal Growth Factor Receptor human, buffered aqueous glycerol solution, 5,000-30,000 units/mg protein (Lowry)
E3138 Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone F4, ascites fluid
E2645 Epidermal Growth Factor Receptor human, lyophilized powder, ≥15,000 units/mg protein (Bradford)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?