• USA Home
  • E1026 - Monoclonal Anti-EphA2 (Eck, Sek-2) antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
E1026 Sigma

Monoclonal Anti-EphA2 (Eck, Sek-2) antibody produced in mouse

clone D7, purified immunoglobulin, buffered aqueous glycerol solution

Synonym: Anti-Eck, Anti-Sek-2

Purchase

Properties

Related Categories Alphabetical Index, Angiogenesis, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology,
species reactivity   rat, bovine, canine (predicted), human, mouse
application(s)   immunohistochemistry: suitable
  immunoprecipitation: suitable
  western blot: 0.5-2 μg/mL using RIPA lysates from human A431 and foreskin fibroblasts, murine 3T3 and rat L6 cells
clone   D7, monoclonal
antibody form   purified immunoglobulin
form   buffered aqueous glycerol solution
isotype   IgG1
mol wt   antigen mol wt 140 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... EPHA2(1969)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

native protein isolated by purification of phosphotyrosine-containing proteins.

General description

The antibody may be used in protein kinase assay.

Physical form

Solution in 0.1 M Tris-glycine, pH 7.4, containing 0.15 M NaCl, 30% glycerol, and 0.05% sodium azide.

Preparation Note

Purified from ascites fluid using protein G.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

EPH receptor A2 is a protein-tyrosine kinase family that belongs to the ephrin receptor subfamily. It binds the ephrin-A and ephrin-B ligands and regulates developmental events, particularly those of the nervous system. EphA2 has been implicated in tumorigenicity of stem-like tumor-propagating cells in human glioblastomas, angiogenesis in Ewing sarcoma cells and cancer metastasis.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Documents

Certificate of Analysis

Certificate of Origin

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?