EMAIL THIS PAGE TO A FRIEND
E111 Sigma

(S)-ENBA

Synonym: (2S)-N6-[2-endo-Norbornyl]adenosine, PD 126,280

  • Empirical Formula (Hill Notation) C17H23N5O4

  • Molecular Weight 361.40

  •  MDL number MFCD00069271

  •  PubChem Substance ID 24278429

Purchase

Properties

Related Categories Adenosines/P2 Nucleotide Receptors (Purinergics), Agonists, Cell Biology, Cell Signaling and Neuroscience, Neuroscience,
InChI Key   YKPCEENRZZBDMC-XSMNFLGNSA-N
solubility   H2O: insoluble0.01 mg/mL
  45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: soluble3 mg/mL
  ethanol, DMSO: soluble
Gene Information   human ... ADORA1(134)

Description

Biochem/physiol Actions

Selective A1 adenosine receptor agonist.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?