• USA Home
  • E1138 - Horizontal submarine electrophoresis units
EMAIL THIS PAGE TO A FRIEND
E1138 Sigma
Horizontal submarine electrophoresis units
gel size 20 cm × 20 cm, base unit, dual mode wide gel 20 cm wide
Purchase
Properties
Related Categories Electrophoresis Supplies, Horizontal Gel Electrophoresis Supplies, Labware, Units and Components for Horizontal Gel Electrophoresis
material   acrylic
gel size   20 cm × 20 cm
Description
General description
Submarine gels may be formed directly on the UV-transparent gel bed using small sliding baffles to contain the agarose as it cools, or outside the unit using a UV-transparent gel tray sealed with tape. Gel is 7.5 cm wide × 10 cm long. Different colors are offered for any color-coding your lab requires (e.g., RNA only).
Combs and tape are not included. Gel tray is not included with the Mini-Gel unit.
Base unit, safety cover, power leads, and all other necessary parts are included.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?