• USA Home
  • E1276 - Monoclonal Anti-Estrogen Receptor β antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
E1276 Sigma

Monoclonal Anti-Estrogen Receptor β antibody produced in mouse

clone 9.88, ascites fluid

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Hormones, Antibodies to Steroid Hormones and Related Products,
species reactivity   human, mouse, rat
application(s)   immunoprecipitation: suitable
  western blot: 1:1,000 using recombinant human estrogen receptor beta
clone   9.88, monoclonal
antibody form   ascites fluid
isotype   IgM
mol wt   antigen mol wt 53 kDa
contains   15 mM sodium azide
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... ESR2(2100)
mouse ... Esr2(13983)
rat ... Esr2(25149)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

recombinant human estrogen receptor β (ER-β).

General description

The antibody recognizes estrogen receptor β and does not cross-react with estrogen receptor α.

Antigen Background

Estrogen Receptor β encodes a member of the family of estrogen receptors and superfamily of nuclear receptor ranscription factors. The gene product contains an N-terminal DNA binding domain and C-terminal ligand binding domain and is localized to the

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)
Western Blotting (1 paper)
Western blotting following immunoprecipitation (1 paper)

Monoclonal Anti-Estrogen Receptor β antibody produced in mouse was used in ELISA of PC12 cells at a working dilution of 1:1500. A dilution of 1:2000 was used in immunoblotting of PC12 whole cell lysates.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

Estrogen-Related Receptors (ERR) are hormone receptors that bind estrogen response elements that in turn bind specific DNA sequences and regulate transcription. Three ERRs are known, alpha, beta and gamma, with highly conserved ligand and DNA binding sequences. ERRβ is selectively expressed in brain and is an important modulator of energy balance, food intake, neuropeptide levels and response to stress.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  nwg
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?