• USA Home
  • E1380 - Electrophoresis film for agarose gels

EMAIL THIS PAGE TO A FRIEND
E1380 Sigma

Electrophoresis film for agarose gels

sheet size 100 mm × 150 mm

Purchase

Properties

description   Gels 7 mil (0.2 mm) Thickness
sheet size   100 mm × 150 mm

Description

Application

Electrophoresis film is a flexible, transparent polyester film specifically designed to support agarose gels. Ideal for casting horizontal agarose gels for immunoelectrophoresis and immunodiffusion. One side has been treated with a hydrophilic coating, which enhances adhesion with aqueous agarose gel media.

Caution

Fluorescence may be exhibited under ultraviolet light which may interfere with some fluorimetric detection methods.

General description

• Gels cast on the electrophoresis film remain permanently attached to the film during electrophoresis, staining and drying
• Handling of thin agarose gels is much easier when attached to electrophoresis film
• Gels attached to electrophoresis film remain flexible after drying

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

E0264 Electrophoresis film for agarose gels, sheet W × L 85 mm × 100 mm

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?