• USA Home
  • E1653 - Estrogen Receptor-β human

EMAIL THIS PAGE TO A FRIEND
E1653 Sigma

Estrogen Receptor-β human

≥80% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution

Purchase

Properties

Related Categories Cancer Research, Cell Biology, Cell Signaling and Neuroscience, Cytokines, Growth Factors and Hormones, Estrogen,
recombinant   expressed in baculovirus infected insect cells
assay   ≥80% (SDS-PAGE)
form   buffered aqueous glycerol solution
mol wt   mol wt ~53.4 kDa
storage temp.   −70°C
Gene Information   human ... ESR2(2100)

Description

Physical form

Solution in 50 mM BIS-TRIS-Propane, pH 9.0, 400 mM KCl, 2 mM DTT, 1 mM EDTA ,and 10% glycerol.

Application

Hormone-inducible transcription factor. Shows high homology with ER-α, tested for its ability to bind both [3H] estradiol and fluorescent estrogens.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?