• USA Home
  • E1889 - (+)-Pseudoephedrine hydrochloride

EMAIL THIS PAGE TO A FRIEND
E1889 Sigma

(+)-Pseudoephedrine hydrochloride

Sigma Reference Standard

Synonym: (+)-ψ-Ephedrine hydrochloride, (1S,2S)-2-Methylamino-1-phenylpropanol hydrochloride, d-Isoephedrine hydrochloride, d-Pseudoephedrine hydrochloride

Purchase

Properties

Related Categories Alkaloid, Cell Biology, Nutrition Research, Phytochemicals by Chemical Classification
grade   Sigma Reference Standard
InChI Key   BALXUFOVQVENIU-KXNXZCPBSA-N
form   neat
packaging   vial of 250 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   185-188 °C(lit.)
format   neat

Description

Biochem/physiol Actions

Non-selective adrenergic agonist; decongestant

Packaging

Supplied in amber screw-cap vials

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Pseudoephedrine Enantiomers on Astec® CHIROBIOTIC® T

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Pseudoephedrine Enantiomers on Astec® CHIROBIOTIC® T
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?