• USA Home
  • E2020 - Express-Ion exchanger C sodium form

EMAIL THIS PAGE TO A FRIEND
E2020 Sigma

Express-Ion exchanger C sodium form

microgranular, 60-130 μm

Purchase

Properties

Related Categories Cation Exchange Media, Ion Exchange Chromatography, Molecular Biology, Protein Chromatography, Proteomics More...
form   microgranular
feature   flow rate 150 cm/hr
matrix active group   carboxymethyl
particle size   60-130 μm
pH range   4.5 - 10
capacity   1 meq/g

Description

Legal Information

Express-Ion is a trademark of Whatman International Ltd.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?