• USA Home
  • E2139 - Ecgonine-d3 methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
E2139 Sigma

Ecgonine-d3 methyl ester hydrochloride

Purchase

Properties

Related Categories Alkaloids / Nitrogen Containing Compounds, Analytical Standards, Analytical/Chromatography, Phytochemical Standards
form   neat
drug control   Home Office Schedule 2; psychotroperegulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?