• USA Home
  • E2252 - Eriochrome® Blue Black B

EMAIL THIS PAGE TO A FRIEND
E2252 Sigma

Eriochrome® Blue Black B

Synonym: Mordant Black 3

  •  Colour Index Number 14640

  •  EC Number 222-639-6

Purchase

Properties

composition   Dye content, ~50%

Description

Legal Information

Eriochrome is a registered trademark of Huntsman Petrochemical, LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
QK2195000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?