• USA Home
  • E2270 - Sulfoxyethyl cellulose

EMAIL THIS PAGE TO A FRIEND
E2270 Sigma

Sulfoxyethyl cellulose

microgranular, 60-130 μm

Purchase

Properties

form   microgranular
feature   flow rate 150 cm/hr
particle size   60-130 μm
pH range   2 - 12
capacity   1 meq/g

Description

Application

Express-Ion Exchanger S

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

S4027 Sulfoxyethyl cellulose Fast Flow, fibers

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?