• USA Home
  • E2278 - E-Cadherin/Fc Chimera human

EMAIL THIS PAGE TO A FRIEND
E2278 Sigma

E-Cadherin/Fc Chimera human

≥90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym: L-CAM, Arc-1, Cell-CAM120/80, Epithelial cadherin, Uvomorulin

Purchase

Description

Physical form

Lyophilized from a 0.2 μm filtered solution in 50 mM Tris-Citrate, 250 mM NaCl, and 2 mM CaCl2 pH 6.5.

Biochem/physiol Actions

E (epithelial)-cadherin is involved in hemophilic cell-cell adhesion in a calcium-dependent manner. It forms adheren junctions through its ectodomains, to maintain physical contacts between adjacent cells. It plays essential roles in developmental processes, tissue homeostasis and multiple diseases. In humans, this protein might be involved in the pathophysiology of ulcerative colitis. In Crohn′s dieases patients, mutations in this gene lead to a truncated or mislocalized protein product. Epithelial-mesenchymal transition (EMT) and mesenchymal-epithelial transition (MET) are the underlying processes of tumorigenesis and embryogenesis, and these involve the control of E-cadherin expression.

General description

E (epithelial)-cadherin is a type I classical cadherin, which exists as a dimer. It has an ectodomain made of five extracellular cadherin-like (EC) domains, connected by conserved linker regions, which are calcium-binding in nature. It spans the membrane once, and contains a cytoplasmic region. It is a glycoprotein, and in humans has a molecular weight of 120kDa. It binds to catenin and cytoskeletal protein with its C-terminal domain.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?