• USA Home
  • E2384 - Electric organ acetone powder from Electrophorus electricus (electric eel)

EMAIL THIS PAGE TO A FRIEND
E2384 Sigma

Electric organ acetone powder from Electrophorus electricus (electric eel)

Purchase

Properties

Related Categories Acetone Powders, Biochemicals and Reagents, Proteins and Derivatives More...
storage temp.   −20°C

Description

Application

Source of electric eel (Electrophorus electricus) organ proteins; contains acetylcholinesterase activity.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?