• USA Home
  • E2391 - Endo-α-N-acetylgalactosaminidase from Diplococcus pneumoniae

EMAIL THIS PAGE TO A FRIEND
E2391 Sigma

Endo-α-N-acetylgalactosaminidase from Diplococcus pneumoniae

buffered aqueous glycerol solution

Purchase

Properties

form   buffered aqueous glycerol solution
impurities   BSA, free
storage temp.   2-8°C

Description

Unit Definition

One unit will catalyze the release of 1 μmole of Gal-GalNAc from native asialofetuin per min at pH 6.0 at 37°C.

Physical form

Solution in 25% glycerol containing 10 mM sodium cacodylate, pH 6.0

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

G1163 O-Glycosidase from Streptococcus pneumoniae, recombinant, expressed in E. coli, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?