• USA Home
  • E2885 - Anti-17β-Estradiol antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
E2885 Sigma

Anti-17β-Estradiol antibody produced in rabbit

whole antiserum, lyophilized powder

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Hormones, Antibodies to Steroid Hormones and Related Products,
biological source   rabbit
antibody form   whole antiserum
clone   polyclonal
form   lyophilized powder
application(s)   radioimmunoassay: suitable
conjugate   unconjugated
storage temp.   2-8°C

Description

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Immunogen

17β-Estradiol-6-BSA

Reconstitution

Reconstitute with 5 ml buffer: 0.05 M Tris-HCl, pH 8.0, 0.1 M sodium chloride, 0.1% gelatin and 15 mM sodium azide. Store this stock at −20 °C. Working dilution (1:10) to be discarded if not used within 12 hours.

Application

Anti-17β-Estradiol antibody may be used for RIA. The antibody has also been used for ELISA.

General description

17β-estradiol (E2) is a sex hormone that is a key regulator of growth, differentiation and function of reproductive tissues, skeletal and cardiovascular systems. The effect of 17β-estradiol is mediated by two intracellular receptors, ERα and ERβ. Although the two receptors interact with similar ligands, have identical DNA-binding domains and have redundant roles in estrogen signalling, they show distinct expression pattern. On binding to the receptor, the estrogen-ER complex enters nucleus and regulates the transcription of specific genes. The biological effects of estradiol may be mediated by the classic binding of estrogen and ER, by upstream activation of the pathway by growth factors. Some pathways that activate the estrogen-ER pathway are those mediated by AP-1, IGF-1, EGF, MAPK and PI3K/Akt.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?