• USA Home
  • E3000 - (−)-Ephedrine Anhydrous

EMAIL THIS PAGE TO A FRIEND
E3000 Sigma

(−)-Ephedrine Anhydrous

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
KB0700000
Flash Point(F) 
186.8 °F
Flash Point(C) 
86 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Ephedrine Enantiomers on CHIRALPAK® AGP

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Ephedrine Enantiomers on CHIRALPAK® AGP
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

Related Products

replaced by

Product #

Description

Add to Cart

134910 (1R,2S)-(−)-Ephedrine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?