• USA Home
  • E3005 - des-Tyr1-γ-Endorphin

EMAIL THIS PAGE TO A FRIEND
E3005 Sigma

des-Tyr1-γ-Endorphin

≥97% (HPLC)

Synonym: β-Lipotropin 62-77

  • CAS Number 67810-56-4

  • Empirical Formula (Hill Notation) C74H122N18O25S

  • Molecular Weight 1695.93

  •  MDL number MFCD00076397

  •  PubChem Substance ID 24894492

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Endorphins, Opioid Peptides, Peptides and Proteins,
InChI Key   YQEWLSJJOSZFHG-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   human ... POMC(5443)

Description

Amino Acid Sequence

Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-Leu

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?