• USA Home
  • E3135 - Anti-Estrone antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
E3135 Sigma

Anti-Estrone antibody produced in rabbit

whole antiserum, lyophilized powder

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Hormones, Antibodies to Steroid Hormones and Related Products,
biological source   rabbit
antibody form   whole antiserum
clone   polyclonal
form   lyophilized powder
application(s)   radioimmunoassay: suitable
conjugate   unconjugated
storage temp.   2-8°C

Description

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Immunogen

estrone-6-thyroglobulin.

Reconstitution

Reconstitute with 5 mL buffer: 0.05 M Tris-HCl, pH 8.0, 0.1 M sodium chloride, 0.1% gelatin and 15 mM sodium azide. Working dilution (1:10) to be discarded if unused within 12 hours.

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?